Learn More
MSC Sellstrom Replacement Window, Acetate, Anti-Fog Coating, Clear Tint, 24/Case
Catalog No. 5000767928
Encompass
This replacement window is designed for the 390 Series Safety Face Shields. It is made from acetate material and features an anti-fog coating, providing clear vision during use. The window has a clear tint.
  • Anti-fog coating helps maintain clear visibility in various conditions.
  • Clear tint ensures unobstructed view.
  • Replacement part extends the life of 390 Series Safety Face Shields.
Catalog No. 50-007-67928 Supplier MSC Supplier No. 78085958
This replacement window is designed for the 390 Series Safety Face Shields. It is made from acetate material and features an anti-fog coating, providing clear vision during use. The window has a clear tint.
  • Anti-fog coating helps maintain clear visibility in various conditions.
  • Clear tint ensures unobstructed view.
  • Replacement part extends the life of 390 Series Safety Face Shields.