Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selnoflast | 2260969-36-4 | MFCD34579587 | 99.5% | 391.53 g/mol | C20H29N3O3S | 5 MG
SDP

Catalog No. 5000262195
Encompass_Preferred

Selnoflast is an orally active, selective, and reversible small-molecule inhibitor of the NLRP3 inflammasome, developed for research into inflammatory pathways and disease models. It is supplied for research use and is characterized by analytical data to support reproducible experimental use.

  • Selective inhibitor of the NLRP3 inflammasome.
  • Orally active and reversible small-molecule mechanism.
  • Suitable for in vitro and in vivo inflammatory disease research.
  • High purity supporting analytical consistency.
  • Soluble in DMSO for standard assay formulations.
  • Molecular weight 391.53 g/mol; molecular formula C20H29N3O3S.

Catalog No. 50-002-62195 Supplier Medchemexpress LLC Supplier No. HY1328315MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selnoflast is an orally active, selective, and reversible small-molecule inhibitor of the NLRP3 inflammasome, developed for research into inflammatory pathways and disease models. It is supplied for research use and is characterized by analytical data to support reproducible experimental use.

  • Selective inhibitor of the NLRP3 inflammasome.
  • Orally active and reversible small-molecule mechanism.
  • Suitable for in vitro and in vivo inflammatory disease research.
  • High purity supporting analytical consistency.
  • Soluble in DMSO for standard assay formulations.
  • Molecular weight 391.53 g/mol; molecular formula C20H29N3O3S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.