Learn More
Cayman Chemical SelonsertIb 10mg
SDP
Catalog No. 502983132
Encompass_Preferred
An ASK1 inhibitor (IC50 = 5.01 nM); decreases apoptosis and stellate cell activation in an in vitro 3D-human liver microtissue model of palmitic acid-induced NASH at 1 µM; sensitizes KB-C2 ABCB1-expressing epidermoid carcinoma cells to doxorubicin and paclitaxel; reduces hepatic necrosis and increases in serum TNF-α, IL-6, and CCL2 in a mouse model of LPS/D-galactosamine-induced acute liver failure at 15, 30, or 60 mg/kg
Catalog No. 50-298-3132 Supplier Cayman Chemical Supplier No. 2097210 MG
An ASK1 inhibitor (IC50 = 5.01 nM); decreases apoptosis and stellate cell activation in an in vitro 3D-human liver microtissue model of palmitic acid-induced NASH at 1 µM; sensitizes KB-C2 ABCB1-expressing epidermoid carcinoma cells to doxorubicin and paclitaxel; reduces hepatic necrosis and increases in serum TNF-α, IL-6, and CCL2 in a mouse model of LPS/D-galactosamine-induced acute liver failure at 15, 30, or 60 mg/kg