Learn More
Sino Biological SELP / P Selectin Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 13025R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SELP / P Selectin (rh SELP / P Selectin; Catalog#13025-H08H; AAN06828.1; Met1-Ala771).
Catalog No. 50-224-8182