Learn More
Medchemexpress LLC HY-114370 10mg , Selpercatinib CAS:2152628-33-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11437010MG
Encompass_Preferred
HY-114370 10mg Selpercatinib CAS: 2152628-33-4 Selpercatinib (LOXO-292) is a potent, selective RET kinase inhibitor with IC50 values of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M), and RET (G810R), respectively. Selpercatinib has anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9318