Learn More
Medchemexpress LLC HY-114370 5mg Medchemexpress, Selpercatinib CAS:2152628-33-4 Purity:>98%
SDP
Catalog No. 501969337
Encompass_Preferred
Medchemexpress, HY-114370 5mg Selpercatinib CAS:2152628-33-4 Selpercatinib (LOXO-292) is a RET kinase inhibitor extracted from patent WO2018071447A1, Compound Example 163, has an IC50 of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M) , and RET (G810R), respectively. Antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9337 Supplier Medchemexpress LLC Supplier No. HY1143705MG
Medchemexpress, HY-114370 5mg Selpercatinib CAS:2152628-33-4 Selpercatinib (LOXO-292) is a RET kinase inhibitor extracted from patent WO2018071447A1, Compound Example 163, has an IC50 of 14.0 nM, 24.1 nM, and 530.7 nM for RET (WT), RET (V804M) , and RET (G810R), respectively. Antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.