Learn More
Aviva Systems Biology Corp SELENOS Antibody - middle region (ARP48082_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48082_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SELS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SELS. Alternate names: AD-015, ADO15, MGC104346, MGC2553, SBBI8, SEPS1, VIMP, SELSApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59714