Learn More
Medchemexpress LLC Seltorexant | 1293281-49-8 | 99.8% | 407.44 | 10 MG
SDP
Catalog No. 5000346001
Encompass_Preferred
Seltorexant (JNJ-42847922) is an orally active, high-affinity, and selective orexin-2 receptor (OX2R) antagonist. It crosses the blood-brain barrier and quickly occupies OX2R binding sites in the rat brain.
  • Orally active
  • High-affinity and selective orexin-2 receptor (OX2R) antagonist
  • Crosses the blood-brain barrier
  • Quickly occupies OX2R binding sites in the rat brain
  • Dose-dependently induces and prolongs sleep in male Sprague-Dawley rats
  • Sleep-promoting effects maintained upon 7-day repeated dosing in rats
Catalog No. 50-003-46001 Supplier Medchemexpress LLC Supplier No. HY10901250MG
Seltorexant (JNJ-42847922) is an orally active, high-affinity, and selective orexin-2 receptor (OX2R) antagonist. It crosses the blood-brain barrier and quickly occupies OX2R binding sites in the rat brain.
  • Orally active
  • High-affinity and selective orexin-2 receptor (OX2R) antagonist
  • Crosses the blood-brain barrier
  • Quickly occupies OX2R binding sites in the rat brain
  • Dose-dependently induces and prolongs sleep in male Sprague-Dawley rats
  • Sleep-promoting effects maintained upon 7-day repeated dosing in rats