Learn More
Medchemexpress LLC HY-50706 50mg Medchemexpress, Selumetinib CAS:606143-52-6 Purity:>98%
SDP
Catalog No. 502029330
Encompass_Preferred
Medchemexpress, HY-50706 50mg Selumetinib CAS:606143-52-6 Selumetinib (AZD6244) is selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. Selumetinib (AZD6244) inhibits ERK1/2 phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9330 Supplier Medchemexpress LLC Supplier No. HY5070650MG
Medchemexpress, HY-50706 50mg Selumetinib CAS:606143-52-6 Selumetinib (AZD6244) is selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. Selumetinib (AZD6244) inhibits ERK1/2 phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.