Learn More
Selleck Chemical LLC Selumetinib sulfate E6024-50MG
SDP
Catalog No. 5000041709
Encompass_Preferred
Selumetinib sulfate (AZD6244 sulfate ARRY-142886 sulfate) is an oral non-ATP-competitive inhibitor of MEK1/2 with an IC50 of 14 nmol/L against purified MEK1 It exhibits an anti-tumor effect in the HT-29 xenograft model and is used to treat cancer and inflammation
Catalog No. 50-000-41709 Supplier Selleck Chemical LLC Supplier No. E602450MG
Selumetinib sulfate (AZD6244 sulfate ARRY-142886 sulfate) is an oral non-ATP-competitive inhibitor of MEK1/2 with an IC50 of 14 nmol/L against purified MEK1 It exhibits an anti-tumor effect in the HT-29 xenograft model and is used to treat cancer and inflammation