Learn More
Medchemexpress LLC Selumetinib sulfate | 943332-08-9 | 99.58% | C17H17BrClFN4O7S | 1 G
SDP
Catalog No. 5000358066
Encompass_Preferred
Selumetinib sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier (BBB).
  • Causes a time- and dose-dependent reduction in DNA synthesis and cell viability
  • Induces growth arrest and apoptosis associated with the inactivation of ERK in primary cells
  • Shows anti-proliferative effects through G0/G1 arrest
  • Results in growth inhibition of cell lines containing B-Raf and Ras mutations
  • Decreases the growth rate of xenografts in a dose-dependent manner
  • Suppresses the growth of xenografts
  • Inhibits ERK1/2 phosphorylation and growth of xenograft tumors in nude mice
  • Tumor regressions observed in xenograft models
Catalog No. 50-003-58066 Supplier Medchemexpress LLC Supplier No. HY50706A1G
Selumetinib sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier (BBB).
  • Causes a time- and dose-dependent reduction in DNA synthesis and cell viability
  • Induces growth arrest and apoptosis associated with the inactivation of ERK in primary cells
  • Shows anti-proliferative effects through G0/G1 arrest
  • Results in growth inhibition of cell lines containing B-Raf and Ras mutations
  • Decreases the growth rate of xenografts in a dose-dependent manner
  • Suppresses the growth of xenografts
  • Inhibits ERK1/2 phosphorylation and growth of xenograft tumors in nude mice
  • Tumor regressions observed in xenograft models