Learn More
Medchemexpress LLC Selumetinib (sulfate) | 943332-08-9 | 99.58% | C17H17BrClFN4O7S | 100 MG
SDP
Catalog No. 5000358352
Encompass_Preferred
Selumetinib (AZD6244) sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier.
  • Selective, non-ATP-competitive oral MEK1/2 inhibitor
  • Potency: Exhibits an IC50 of 14 nM for MEK1
  • Inhibits ERK1/2 phosphorylation
  • Capable of penetrating the blood-brain barrier
  • Causes time- and dose-dependent reduction in DNA synthesis and cell viability
  • Induces growth arrest and apoptosis
  • Demonstrates anti-proliferative effects through G0/G1 arrest
  • Leads to growth inhibition in various cell lines with B-Raf and Ras mutations
  • Decreases growth rate of xenografts
  • Induces tumor regressions
Catalog No. 50-003-58352 Supplier Medchemexpress LLC Supplier No. HY50706A100MG
Selumetinib (AZD6244) sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier.
  • Selective, non-ATP-competitive oral MEK1/2 inhibitor
  • Potency: Exhibits an IC50 of 14 nM for MEK1
  • Inhibits ERK1/2 phosphorylation
  • Capable of penetrating the blood-brain barrier
  • Causes time- and dose-dependent reduction in DNA synthesis and cell viability
  • Induces growth arrest and apoptosis
  • Demonstrates anti-proliferative effects through G0/G1 arrest
  • Leads to growth inhibition in various cell lines with B-Raf and Ras mutations
  • Decreases growth rate of xenografts
  • Induces tumor regressions