Learn More
Medchemexpress LLC Selumetinib (sulfate) | 943332-08-9 | C17H17BrClFN4O7S | 200 MG
SDP
Catalog No. 5000358272
Encompass_Preferred
Selumetinib (AZD6244) sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier (BBB).
  • Inhibits ERK1/2 phosphorylation
  • Penetrates the blood-brain barrier
  • Reduces DNA synthesis and cell viability
  • Induces growth arrest and apoptosis
  • Shows anti-proliferative effects through G0/G1 arrest
  • Inhibits growth of cell lines with B-Raf and Ras mutations
  • Decreases growth rate of xenografts
  • Causes tumor regressions in xenograft models
Catalog No. 50-003-58272 Supplier Medchemexpress LLC Supplier No. HY50706A200MG
Add to Cart
Add to Cart
Selumetinib (AZD6244) sulfate is a selective, non-ATP-competitive oral MEK1/2 inhibitor, with an IC50 of 14 nM for MEK1. It inhibits ERK1/2 phosphorylation and can penetrate the blood-brain barrier (BBB).
  • Inhibits ERK1/2 phosphorylation
  • Penetrates the blood-brain barrier
  • Reduces DNA synthesis and cell viability
  • Induces growth arrest and apoptosis
  • Shows anti-proliferative effects through G0/G1 arrest
  • Inhibits growth of cell lines with B-Raf and Ras mutations
  • Decreases growth rate of xenografts
  • Causes tumor regressions in xenograft models