Learn More
Medchemexpress LLC Selvigaltin (GB1211) | 1978336-95-6 | 99.9% | 533.32 | 25 MG
SDP
Catalog No. 5000363467
Encompass_Preferred
Selvigaltin (GB1211) is an orally active small molecule inhibitor of galectin-3. It demonstrates anti-tumor activity and is effective at reducing galectin-3 levels in the liver. It also decreases biomarkers of liver function, inflammation, and fibrosis, while restoring T-cell activity.
  • Reduces galectin-3 levels in the liver
  • Decreases biomarkers of liver function (AST, ALT, bilirubin)
  • Lowers inflammation markers (cells foci)
  • Reduces fibrosis biomarkers (PSR, SHG)
  • Decreases mRNA and protein expression of IL6, TGFβ3, SNAI2, and collagen
  • Restores T-cell activity
  • Leads to fewer tumors and metastasis
Catalog No. 50-003-63467 Supplier Medchemexpress LLC Supplier No. 25MGHY147041
Selvigaltin (GB1211) is an orally active small molecule inhibitor of galectin-3. It demonstrates anti-tumor activity and is effective at reducing galectin-3 levels in the liver. It also decreases biomarkers of liver function, inflammation, and fibrosis, while restoring T-cell activity.
  • Reduces galectin-3 levels in the liver
  • Decreases biomarkers of liver function (AST, ALT, bilirubin)
  • Lowers inflammation markers (cells foci)
  • Reduces fibrosis biomarkers (PSR, SHG)
  • Decreases mRNA and protein expression of IL6, TGFβ3, SNAI2, and collagen
  • Restores T-cell activity
  • Leads to fewer tumors and metastasis