Learn More
Electron Microscopy Sciences SEM Aluminum Tungsten Dendrites
SDP
Catalog No. 5027095
Encompass_Preferred
The various spacings created by the dendritic structure give the gap test, and the topographical arrangement of the dendrites leads to the gray level test. The specimen is non-magnetic, vacuum clean. It is most useful for working in the probe size range of 25 to 75 nm. Supplied unmounted.
Catalog No. 50-270-95 Supplier Electron Microscopy Sciences Supplier No. 7951401
The various spacings created by the dendritic structure give the gap test, and the topographical arrangement of the dendrites leads to the gray level test. The specimen is non-magnetic, vacuum clean. It is most useful for working in the probe size range of 25 to 75 nm. Supplied unmounted.