Learn More
Electron Microscopy Sciences SEM Clip M4 Cylinder Mount, 3 clips, 25 mm, 6 mm Pin height
SDP
Catalog No. 501927120
Encompass_Preferred
These spring-loaded clips accommodate samples of up to 2 mm thickness. Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Available with single and multiple clips and are ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal and more. SEM Clip Mounts with multiple clips can hold larger samples or multiple smaller samples.
Includes: 3 clips
Diameter: 25 mm
Pin height: 6 mm
Catalog No. 50-192-7120 Supplier Electron Microscopy Sciences Supplier No. 7593020
These spring-loaded clips accommodate samples of up to 2 mm thickness. Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Available with single and multiple clips and are ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal and more. SEM Clip Mounts with multiple clips can hold larger samples or multiple smaller samples.
Includes: 3 clips
Diameter: 25 mm
Pin height: 6 mm