Learn More
Electron Microscopy Sciences Clip SEM Pin Mount, 38 mm, 1 clip
SDP
Catalog No. 501927111
Encompass_Preferred
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs. These spring-loaded clips accommodate samples of up to 2 mm thickness. Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, etc.
Diameter: 38 mm
Includes: 1 clip
Catalog No. 50-192-7111 Supplier Electron Microscopy Sciences Supplier No. 7592318
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs. These spring-loaded clips accommodate samples of up to 2 mm thickness. Samples are held securely by small spring-loaded clips and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time. Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, etc.
Diameter: 38 mm
Includes: 1 clip