Learn More
Mopec SEM Collection Devices
SDP
Catalog No. 22446326 Shop All Mopec Products
Change view
Click to view available options
For Use With (Application):
D-Stub SEM Collection
Q-Stub SEM Collection
Standard SEM Collection
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
22-446-326 Standard SEM Collection
22-446-325 Q-Stub SEM Collection
22-446-269 D-Stub SEM Collection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-446-326 Supplier Mopec Supplier No. FS484
Add to Cart
Add to Cart
Used to collect gunshot residue and other trace evidence for SEM analysis, each vial contains an adhesive carbon stub and a write-on label for sample identification. Multiple stub formats are available to support different collection requirements while maintaining organized sample handling.
Construction
  • Each vial contains a SEM stub covered with a double-sided adhesive carbon tape
  • Write-on label
Performance
  • For collection of explosive residue collection, inorganic particle matter, preparing standards for future comparison, paint chips, glass, soil, fiber, or hair
Materials
  • Plastic
  • Carbon tape
Quantity
  • 50 per case
Specifications
For Use With (Application) Standard SEM Collection
Quantity 50/Cs.