Learn More
Electron Microscopy Sciences Scanning Electron Microscope Finder Grid, Nickel
SDP
Catalog No. 5028592
Encompass_Preferred
Finder grids for SEM; a valuable tool for analytical studies. The grids are available in 10 mm diameter and they can be placed directly on the stub surface and used to identify the area of interest in which the studies are performed.
Catalog No. 50-285-92 Supplier Electron Microscopy Sciences Supplier No. 80100NI
Finder grids for SEM; a valuable tool for analytical studies. The grids are available in 10 mm diameter and they can be placed directly on the stub surface and used to identify the area of interest in which the studies are performed.