Learn More
Electron Microscopy Sciences SEM Finder Grid SEMF1, Nickel
SDP
Catalog No. 5028768
Encompass_Preferred
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. Four quadrant markers tapered towards center. 100 radial sectors.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm
Catalog No. 50-287-68 Supplier Electron Microscopy Sciences Supplier No. 80101NI
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. Four quadrant markers tapered towards center. 100 radial sectors.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm