Learn More
Electron Microscopy Sciences SEM Finder Grid SEMF2, Nickel
SDP
Catalog No. 5028763
Encompass_Preferred
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. Larger cells identified 1-57 & subdivided into 4. 228 identifiable cells.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm
Catalog No. 50-287-63 Supplier Electron Microscopy Sciences Supplier No. 80102NI
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. Larger cells identified 1-57 & subdivided into 4. 228 identifiable cells.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm