Learn More
Electron Microscopy Sciences SEM Finder Grid SEMF3, Nickel
SDP
Catalog No. 5028765
Encompass_Preferred
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. 25 cells identified by reference to alpha-numeric position.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm
Catalog No. 50-287-65 Supplier Electron Microscopy Sciences Supplier No. 80103NI
Designed to aid in the identification and localization of SEM specimens when placed on standard SEM stubs. 25 cells identified by reference to alpha-numeric position.
Material: Nickel
Diameter: 10 mm
Thickness: ~50 µm