Learn More
Electron Microscopy Sciences SEM FIXATIVE: GLUTARALDEHYDE 2
SDP
Catalog No. NC2029340
Encompass_Preferred
SEM fixative: Glutaraldehyde 2% in 0.1M Sodium Cacodylate Buffer, pH 7.4
Catalog No. NC2029340 Supplier Electron Microscopy Sciences Supplier No. 1653620
SEM fixative: Glutaraldehyde 2% in 0.1M Sodium Cacodylate Buffer, pH 7.4