Learn More
Electron Microscopy Sciences SEM Wafer Holder, 100 mm
SDP
Catalog No. 501928211
Encompass
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 4" (100 mm) diameter wafer.
Size: 100 mm
Catalog No. 50-192-8211 Supplier Electron Microscopy Sciences Supplier No. 7595814
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 4" (100 mm) diameter wafer.
Size: 100 mm