Learn More
Electron Microscopy Sciences SEM Wafer Holder, 150 mm
SDP
Catalog No. 501928212
Encompass
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 5" (150 mm) diameter wafer.
Size: 150 mm
Catalog No. 50-192-8212 Supplier Electron Microscopy Sciences Supplier No. 7595815
These wafer holders are made from aluminum and feature brass clips which secure the wafers. Each holder comes with a 0.125" (3.2 mm) pin of 0.375" (9.5 mm) length. Accomodates 5" (150 mm) diameter wafer.
Size: 150 mm