Learn More
Thermo Scientific SEM Detector
Catalog No. NC0802595 Shop All Thermo Scientific Products
Encompass
SEM Detector for use with iCAP Q/Qnova Series ICP-MS systems
Catalog No. NC0802595 Supplier Thermo Scientific Supplier No. 1299710
SEM Detector for use with iCAP Q/Qnova Series ICP-MS systems