Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Mini, 6.6 x 1.3 mm Head, 9.5 mm Pin, Aluminum
SDP
Catalog No. 501901459
Encompass_Preferred
Mini SEM pin stub mount for Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan, ETEC and Amray. Made from lathe finished ultra-pure aluminum.
Head: 6.6 x 1.3 mm
Pin height: 3/8"
Catalog No. 50-190-1459 Supplier Electron Microscopy Sciences Supplier No. 7563830
Mini SEM pin stub mount for Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, Tescan, ETEC and Amray. Made from lathe finished ultra-pure aluminum.
Head: 6.6 x 1.3 mm
Pin height: 3/8"