Learn More
Electron Microscopy Sciences Hitachi M4 SEM Cylinder Mount 1-1/4" surface with 12 Divisions, 1/4" height, Aluminum
SDP
Catalog No. 501901479
Encompass_Preferred
Hitachi M4 Thread SEM mounts made from lathe finished ultra-pure aluminum.
Head: 1-1/4"
Height: 1/4"
Catalog No. 50-190-1479 Supplier Electron Microscopy Sciences Supplier No. 7563590D
Hitachi M4 Thread SEM mounts made from lathe finished ultra-pure aluminum.
Head: 1-1/4"
Height: 1/4"