Learn More
Electron Microscopy Sciences Hitachi M4 SEM Cylinder Mount 5/8" surface, 3/8" height, Aluminum
SDP
Catalog No. 501901483
Encompass_Preferred
Hitachi M4 Thread SEM mounts made from lathe finished ultra-pure aluminum.
Head: 5/8"
Height: 3/8"
Catalog No. 50-190-1483 Supplier Electron Microscopy Sciences Supplier No. 7563510
Hitachi M4 Thread SEM mounts made from lathe finished ultra-pure aluminum.
Head: 5/8"
Height: 3/8"