Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 2-1/2" Head, 3/8" Pin, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501901453
Encompass_Preferred
Slotted head scanning electron microscope (SEM) mount for use with AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, and Tescan microscopes.
Head: 2.5"
Pin height: 3/8"
Pin diameter: 3.2 mm
Catalog No. 50-190-1453 Supplier Electron Microscopy Sciences Supplier No. 7563690
Slotted head scanning electron microscope (SEM) mount for use with AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/Philips, CamScan, and Tescan microscopes.
Head: 2.5"
Pin height: 3/8"
Pin diameter: 3.2 mm