Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 100 mm Head, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501928129
Encompass
Pin stub mount for use with AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/PHILIPS, CanScan, and Tescan. Made from ultra-pure aluminum with a standard lathe finish.
Head: 100 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-8129 Supplier Electron Microscopy Sciences Supplier No. 7523930
Pin stub mount for use with AMRAY, Cambridge, Leica, ZEISS/LEO, FEI/PHILIPS, CanScan, and Tescan. Made from ultra-pure aluminum with a standard lathe finish.
Head: 100 mm
Pin diameter: 3.2 mm