Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 12 Division 32 mm head with 9 mm Pin, Aluminum, 3.2 mm Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 7518364
Encompass_Preferred
Pin stub mount with 12 divisions for use with FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Made from ultra-pure aluminum with a standard lathe finish.
Head: 32 mm
Pin height: 9 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-8146