Learn More
Electron Microscopy Sciences SEM TM-3000 Angle Mount 25° Angle 25 mm Head, Aluminum
SDP
Catalog No. 501928178
Encompass_Preferred
SEM specimen mount for TM1000/TM3000 with EDS Analysis Systems. These angled mounts feature an orientation cross for precise positioning towards the EDS detector. A tilt provides better detection and higher signal-to-noise ratio for x-ray analysis. Made from Aluminum.
Head: 25 mm
Catalog No. 50-192-8178 Supplier Electron Microscopy Sciences Supplier No. 7531114
SEM specimen mount for TM1000/TM3000 with EDS Analysis Systems. These angled mounts feature an orientation cross for precise positioning towards the EDS detector. A tilt provides better detection and higher signal-to-noise ratio for x-ray analysis. Made from Aluminum.
Head: 25 mm