Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 3-Division 18 mm Head with 9 mm Pin, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501928143
Encompass_Preferred
Pin stub mount with 3 divisions for use with FEI, Phillips, LEO, Zeiss, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Made from ultra-pure aluminum with a standard lathe finish.
Head: 18 mm
Pin height: 9 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-8143 Supplier Electron Microscopy Sciences Supplier No. 7518303
Pin stub mount with 3 divisions for use with FEI, Phillips, LEO, Zeiss, Cambridge, Leica, Amray, Tescan and Camscan SEMs. Made from ultra-pure aluminum with a standard lathe finish.
Head: 18 mm
Pin height: 9 mm
Pin diameter: 3.2 mm