Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 3-Division 1/2" Head with 9 mm Pin, Aluminum, 3.2 mm Pin diameter
SDP
Catalog No. 501901462
Encompass_Preferred
3-Division surface pin stub mounts are for use with FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan are made from lathe finished ultra-pure aluminum.
Head: 1/2"
Pin height: 9 mm
Pin diameter: 3.2 mm
Catalog No. 50-190-1462 Supplier Electron Microscopy Sciences Supplier No. 7518330
3-Division surface pin stub mounts are for use with FEI/Philips, ZEISS/LEO, Cambridge, Leica, Amray, Tescan and Camscan are made from lathe finished ultra-pure aluminum.
Head: 1/2"
Pin height: 9 mm
Pin diameter: 3.2 mm