Learn More
Electron Microscopy Sciences SEM Hitachi M4 45°, 5/8" head, 3/8" height, Aluminum
SDP
Catalog No. 501928186
Encompass_Preferred
45 degree SEM specimen mount with aluminum slotted head for use with Hitachi. Made from ultra-pure aluminum.
Head: 5/8"
Height: 3/8"
Catalog No. 50-192-8186 Supplier Electron Microscopy Sciences Supplier No. 75357
45 degree SEM specimen mount with aluminum slotted head for use with Hitachi. Made from ultra-pure aluminum.
Head: 5/8"
Height: 3/8"