Learn More
Electron Microscopy Sciences Standard Pin SEM Stub 45° Low Profile 1/2" Head with 3/8" Pin, Aluminum
SDP
Catalog No. 501928181
Encompass_Preferred
45° low profile SEM specimen mount for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1/2"
Pin height: 3/8"
Catalog No. 50-192-8181 Supplier Electron Microscopy Sciences Supplier No. 75349
45° low profile SEM specimen mount for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1/2"
Pin height: 3/8"