Learn More
Electron Microscopy Sciences Shorter Pin SEM Stub 45° Low Profile, 1/2" Head with 1/4" Pin, Aluminum
SDP
Catalog No. 501928182
Encompass_Preferred
45° Low Profile SEM stub specimen mount with shorter pin (6 mm) and 45 degree mounting surfaces for use with Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Pin height: 1/4"
Catalog No. 50-192-8182 Supplier Electron Microscopy Sciences Supplier No. 75351
45° Low Profile SEM stub specimen mount with shorter pin (6 mm) and 45 degree mounting surfaces for use with Zeiss/LEO scanning electron microscopes, FE-SEMs & FIB systems. Made from ultra-pure aluminum with a standard lathe finish.
Head: 1/2"
Pin height: 1/4"