Learn More
Electron Microscopy Sciences SEM Cylinder Mount 45° Angle 3/8" Surface, 3.1mm Pin, Aluminum
SDP
Supplier:  Electron Microscopy Sciences 75240
Encompass_Preferred
Scanning electron microscope specimen mount with 45 degree angle. Made from lathe finished ultra-pure aluminum.  
Head: 1/2"
Catalog No. 50-243-48