Learn More
Electron Microscopy Sciences SEM Cylinder Mount 45° Angle 3/8" Surface, Aluminum
SDP
Catalog No. 5024349
Encompass_Preferred
Scanning electron microscope specimen mount with 45 degree angle. Made from lathe finished ultra-pure aluminum.  
Head: 3/8"
Height: 3/8"
Catalog No. 50-243-49 Supplier Electron Microscopy Sciences Supplier No. 75340
Scanning electron microscope specimen mount with 45 degree angle. Made from lathe finished ultra-pure aluminum.  
Head: 3/8"
Height: 3/8"