Learn More
Electron Microscopy Sciences Amray 1000/1200/1400 Pin Stub 1/2" head, Aluminum, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 75140
Encompass_Preferred
Scanning electron microscope specimen mounts for AMRAY 1000/1200/1400 SEMs. Made from ultra-pure aluminum standard lathe finish.
Head: 1/2"
Pin diameter: 1/8"
Catalog No. 50-245-26