Learn More
Electron Microscopy Sciences Amray 1000/1200/1400 Pin Stub 1/2" head, Carbon, 1/8" Pin diameter
SDP
Catalog No. 5032965
Encompass_Preferred
Scanning electron microscope specimen mounts for AMRAY 1000/1200/1400 SEMs. Made from spectroscopically pure carbon.
Head: 1/2"
Pin diameter: 1/8"
Catalog No. 50-329-65 Supplier Electron Microscopy Sciences Supplier No. 76140
Scanning electron microscope specimen mounts for AMRAY 1000/1200/1400 SEMs. Made from spectroscopically pure carbon.
Head: 1/2"
Pin diameter: 1/8"