Learn More
Electron Microscopy Sciences Amray 1000/1200 Pin Stub 1" head, Aluminum, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 75120
Encompass_Preferred
Scanning electron microscope specimen mount for Amray 1000/1200. Made from ultra-pure aluminum with standard lathe finish.  
Head: 1"
Pin diameter: 1/8"
Catalog No. 50-245-23