Learn More
Electron Microscopy Sciences Amray 1400 Pin Stub Slotted 1" head, Carbon, 1/8" Pin diameter
SDP
Supplier:  Electron Microscopy Sciences 76171
Encompass_Preferred
For use with Amray 1400 scanning electron microscopes. Made from spectroscopically pure carbon.
Head: 1/2"
Catalog No. 50-329-69