Learn More
Electron Microscopy Sciences Cambridge S4 Mark II, S-410 Pin Stub 1-1/2" head, 3/16" height, Carbon
SDP
Supplier:  Electron Microscopy Sciences 76180
Encompass_Preferred
Scanning electron microscope specimen mounts for Cambridge S4, Mark II, and S-410. Made from spectroscopically pure carbon.
Head: 1-1/4"
Height: 3/16"
Catalog No. 50-248-47