Learn More
Electron Microscopy Sciences Cambridge S4 Mark II, S-410 Pin Stub 1-1/2" head, 3/16" height, Carbon
SDP
Catalog No. 5024846
Encompass_Preferred
Scanning electron microscope specimen mounts for Cambridge S4, Mark II, and S-410. Made from spectroscopically pure carbon.
Head: 1-1/4"
Height: 3/16"
Catalog No. 50-248-46 Supplier Electron Microscopy Sciences Supplier No. 76181
Scanning electron microscope specimen mounts for Cambridge S4, Mark II, and S-410. Made from spectroscopically pure carbon.
Head: 1-1/4"
Height: 3/16"