Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Slotted 18 mm Head with 8 mm Pin, Carbon, 3.2 mm Pin diameter
SDP
Catalog No. 501928137
Encompass_Preferred
Pin stub mount with slotted head for use with Cambridge, Leica, ZEISS/LEO, FEI/Philips, PHENOM, CamScan, and Tescan.
Head: 18 mm
Pin height: 8 mm
Catalog No. 50-192-8137 Supplier Electron Microscopy Sciences Supplier No. 7563603
Pin stub mount with slotted head for use with Cambridge, Leica, ZEISS/LEO, FEI/Philips, PHENOM, CamScan, and Tescan.
Head: 18 mm
Pin height: 8 mm