Learn More
Electron Microscopy Sciences Coates & Welter SEM Mount 3/4" Head, 3/8 " Pin diameter, Aluminum
SDP
Catalog No. 5025026
Encompass_Preferred
Coates & Welter aluminum specimen mounts for scanning electron microscopes.
Head: 3/4"
Pin diameter: 3/8"
Catalog No. 50-250-26 Supplier Electron Microscopy Sciences Supplier No. 75250
Coates & Welter aluminum specimen mounts for scanning electron microscopes.
Head: 3/4"
Pin diameter: 3/8"