Learn More
Electron Microscopy Sciences Standard Pin SEM Stub Combo 45-90°, 1" Head with 3/8" Pin, Aluminum
SDP
Catalog No. 501928179
Encompass_Preferred
45°/90° SEM specimen mount with aluminum slotted head for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1"
Pin height: 3/8"
Catalog No. 50-192-8179 Supplier Electron Microscopy Sciences Supplier No. 75347
45°/90° SEM specimen mount with aluminum slotted head for use with FEI, Tescan, Zeiss, Philips, LEO, Cambridge, AMRAY, Leica, CamScan, ETEC. Made from ultra-pure aluminum.
Head: 1"
Pin height: 3/8"