Learn More
Electron Microscopy Sciences Combination 45°-90° SEM Pin Mount, Aluminum
SDP
Catalog No. 5033088
Encompass_Preferred
SEM mount with an aluminum slotted head with 3.1 mm diameter pin, fits most SEMs.
Catalog No. 50-330-88 Supplier Electron Microscopy Sciences Supplier No. 75344
SEM mount with an aluminum slotted head with 3.1 mm diameter pin, fits most SEMs.